missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Benzothiophen-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 95%, Thermo Scientific™

Catalog No. CC24839CB
Change view
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC24839CB 250mg Amber Glass Bottle
CC24839DA 1g Amber Glass Bottle
CC24839DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC24839CB Supplier Thermo Scientific Chemicals Supplier No. CC24839CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 501945-71-7
Molecular Formula C14H17BO2S
Molecular Weight (g/mol) 260.158
MDL Number MFCD05664405
InChI Key YFTHTJAPODJVSL-UHFFFAOYSA-N
Synonym 2-benzo b thiophen-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1-benzothiophen-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzothiophene-5-boronic acid pinacol ester, 1-benzothiophen-5-yl-4,4,5,5tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-benzo b thien-5-yl-4,4,5,5-tetramethyl, 1-benzothiophen-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-benzo b thiophen-5-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzo b thiophene, 2-1-benzothiophen-5-yl-4,4,5,5-tetra-methyl-1,3,2-dioxaborolane, benzo b thiophene,4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl
PubChem CID 2795455
IUPAC Name 2-(1-benzothiophen-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)SC=C3

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 260.16
Percent Purity 95%
Chemical Name or Material 2-(1-Benzothiophen-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.