missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester, 97%
CAS: 1073354-07-0 | C11H21BO4 | 228.095 g/mol
$126.05 - $126.05
Chemical Identifiers
| CAS | 1073354-07-0 |
|---|---|
| Molecular Formula | C11H21BO4 |
| Molecular Weight (g/mol) | 228.095 |
| MDL Number | MFCD03788722 |
| InChI Key | DNBRLKJRBDIKOO-UHFFFAOYSA-N |
| Synonym | 2-2-1,3-dioxolan-2-yl ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1,3-dioxolan-2-yl ethylboronic acid pinacol ester, 2-1,3-dioxolan-2-yl-1-ethylboronic acid pinacol ester |
| PubChem CID | 46739008 |
| IUPAC Name | 2-[2-(1,3-dioxolan-2-yl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
| SMILES | B1(OC(C(O1)(C)C)(C)C)CCC2OCCO2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2607103
|
Thermo Scientific Chemicals
H2607103 |
1 g |
Each for $126.05
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1073354-07-0 | |
| 228.095 | |
| DNBRLKJRBDIKOO-UHFFFAOYSA-N | |
| 46739008 | |
| B1(OC(C(O1)(C)C)(C)C)CCC2OCCO2 |
| C11H21BO4 | |
| MFCD03788722 | |
| 2-2-1,3-dioxolan-2-yl ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1,3-dioxolan-2-yl ethylboronic acid pinacol ester, 2-1,3-dioxolan-2-yl-1-ethylboronic acid pinacol ester | |
| 2-[2-(1,3-dioxolan-2-yl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
| 1073354-07-0 | |
| C11H21BO4 | |
| 1 g | |
| DNBRLKJRBDIKOO-UHFFFAOYSA-N | |
| 2-[2-(1,3-dioxolan-2-yl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 46739008 | |
| 97% |
| 100°C (1.5 mmHg) | |
| MFCD03788722 | |
| 2-2-1,3-dioxolan-2-yl ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1,3-dioxolan-2-yl ethylboronic acid pinacol ester, 2-1,3-dioxolan-2-yl-1-ethylboronic acid pinacol ester | |
| B1(OC(C(O1)(C)C)(C)C)CCC2OCCO2 | |
| 228.095 | |
| 228.1 | |
| 2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only