missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzoxadiazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC09701EA
Change view
Click to view available options
Quantity:
250mg
1g
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC09701EA 10g
CC09701CB 250mg
CC09701DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC09701EA Supplier Thermo Scientific Chemicals Supplier No. CC09701EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19155-88-5
Molecular Formula C7H3N2O3
Molecular Weight (g/mol) 163.11
MDL Number MFCD00276977
InChI Key WZUFYJFTOVGJJT-UHFFFAOYSA-M
Synonym benzo c 1,2,5 oxadiazole-5-carboxylic acid, benzofurazan-5-carboxylic acid, 5-benzofurazancarboxylic acid, benzo c 1,2,5-oxadiazole-5-carboxylic acid, pubchem3995, maybridge1_002587, 4-bromo-2,6-dimethylanisole,, 2,1,3-benzoxadiazole-5-carboxylicacid, benzo 1,2,5 oxadiazole-5-carboxylic acid, benzo 2,1,3 oxadiazole-5-carboxylic acid
PubChem CID 2735454
IUPAC Name 2,1,3-benzoxadiazole-5-carboxylic acid
SMILES [O-]C(=O)C1=CC2=NON=C2C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10g
Formula Weight 164.12
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzoxadiazole-5-carboxylic acid