missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzoxadiazol-4-amine, 97%, Thermo Scientific™

Catalog No. BTB13048EA
Change view
Click to view available options
Quantity:
1g
5g
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB13048EA 10g
BTB13048DA 1g
BTB13048DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BTB13048EA Supplier Thermo Scientific Chemicals Supplier No. BTB13048EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 767-63-5
Molecular Formula C6H5N3O
Molecular Weight (g/mol) 135.126
MDL Number MFCD00168448
InChI Key IPCMVRZVNJHUHR-UHFFFAOYSA-N
Synonym 4-aminobenzofurazan, benzo 1,2,5 oxadiazol-4-ylamine, 2,1,3-benzooxadiazol-4-ylamine, benzo c 1,2,5-oxadiazole-4-ylamine, benzofurazan-4-amine, benzofurazane-4-amine, acmc-209p5u, 2,1,3-benzoxadiazol-7-amine, 2,1,3-benzoxadiazol-4-ylamine, benzo c 1,2,5 oxadiazol-4-amine
PubChem CID 584570
IUPAC Name 2,1,3-benzoxadiazol-4-amine
SMILES C1=CC2=NON=C2C(=C1)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10g
Formula Weight 135.13
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzoxadiazol-4-amine