missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzoxadiazol-4-amine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB13048DE

Quantity 5g

 View more versions of this product

Catalog No. BTB13048DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

767-63-5
135.126
IPCMVRZVNJHUHR-UHFFFAOYSA-N
584570
C1=CC2=NON=C2C(=C1)N
C6H5N3O
MFCD00168448
4-aminobenzofurazan, benzo 1,2,5 oxadiazol-4-ylamine, 2,1,3-benzooxadiazol-4-ylamine, benzo c 1,2,5-oxadiazole-4-ylamine, benzofurazan-4-amine, benzofurazane-4-amine, acmc-209p5u, 2,1,3-benzoxadiazol-7-amine, 2,1,3-benzoxadiazol-4-ylamine, benzo c 1,2,5 oxadiazol-4-amine
2,1,3-benzoxadiazol-4-amine

Specifications

2, 1, 3-Benzoxadiazol-4-amine
96.5
97%
5g
4-aminobenzofurazan, benzo 1,2,5 oxadiazol-4-ylamine, 2,1,3-benzooxadiazol-4-ylamine, benzo c 1,2,5-oxadiazole-4-ylamine, benzofurazan-4-amine, benzofurazane-4-amine, acmc-209p5u, 2,1,3-benzoxadiazol-7-amine, 2,1,3-benzoxadiazol-4-ylamine, benzo c 1,2,5 oxadiazol-4-amine
C1=CC2=NON=C2C(=C1)N
135.126
135.13
767-63-5
100.0
C6H5N3O
MFCD00168448
IPCMVRZVNJHUHR-UHFFFAOYSA-N
2,1,3-benzoxadiazol-4-amine
584570
97%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents