missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazole-5-carboxylic acid, 97%

Catalog No. p-7031859
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33095MD 250 mg
AAH3309503 1 g
AAH3309506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH33095MD Supplier Thermo Scientific Chemicals Supplier No. H33095MD
Only null left

CAS: 16405-98-4 | C7H4N2O2S | 180.181 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16405-98-4
Molecular Formula C7H4N2O2S
Molecular Weight (g/mol) 180.181
MDL Number MFCD01647555
InChI Key YHMXJZVGBCACMT-UHFFFAOYSA-N
Synonym benzo c 1,2,5 thiadiazole-5-carboxylic acid, benzo 1,2,5 thiadiazole-5-carboxylic acid, 2??,1,3-benzothiadiazole-5-carboxylic acid, benzo c 1,2,5-thiadiazole-5-carboxylic acid, 4ddm, 2,1,3-benzothiadiazole-5-carboxylicacid, benzo-2,1,3-thiadiazole-5-carboxylic acid, 2,1,3-benzothiadiazole-5-carboxylic acid, benzo 1,2, 5 thiadiazole-5-carboxylic acid, 2$l^ 4 ,1,3-benzothiadiazole-5-carboxylic acid
PubChem CID 602011
IUPAC Name 2,1,3-benzothiadiazole-5-carboxylic acid
SMILES C1=CC2=NSN=C2C=C1C(=O)O

Specifications

Melting Point 228°C to 230°C
Quantity 250 mg
Formula Weight 180.19
Percent Purity 97%
Chemical Name or Material 2,1,3-Benzothiadiazole-5-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only