missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazole-5-carbaldehyde, 97%, Thermo Scientific™

Catalog No. p-2696559
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC09204CB 250mg
CC09204DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC09204CB Supplier Thermo Scientific Chemicals Supplier No. CC09204CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71605-72-6
Molecular Formula C7H4N2OS
Molecular Weight (g/mol) 164.182
MDL Number MFCD00457949
InChI Key GEFIFDVQYCPLHC-UHFFFAOYSA-N
Synonym benzo c 1,2,5 thiadiazole-5-carbaldehyde, 2,1,3-benzothiadiazole-5-carboxaldehyde, benzo 1,2,5 thiadiazole-5-carbaldehyde, 5-formyl-2,1,3-benzothiadiazole, 5-formylbenzo-2,1,3-thiadiazole, benzo 1,2,5 thiadiazol-5-carbaldehyde, benzo 1,2,5 thiadiazole-5-carboxaldehyde
PubChem CID 2776290
IUPAC Name 2,1,3-benzothiadiazole-5-carbaldehyde
SMILES C1=CC2=NSN=C2C=C1C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250mg
Formula Weight 164.19
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzothiadiazole-5-carbaldehyde