missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazole-4-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. CC09103DA
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC09103DA 1g
CC09103DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC09103DA Supplier Thermo Scientific Chemicals Supplier No. CC09103DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 73713-79-8
Molecular Formula C6H3ClN2O2S2
Molecular Weight (g/mol) 234.67
MDL Number MFCD00068049
InChI Key CXAICGCTHOWKPP-UHFFFAOYSA-N
Synonym benzo c 1,2,5 thiadiazole-4-sulfonyl chloride, benzo 2,1,3 thiadiazole-4-sulfonyl chloride, 2,1,3-benzothiadiazole-4-sulphonyl chloride, 2,1,3-benzothiadiazol-4-sulfonyl chloride, 4-chlorosulfonyl-2,1,3-benzothiadiazole, 2,1,3-benzothiadiazole, 4-chlorosulfonyl, 2,1,3-benzothiadiazole-4-sulfonylchloride, benzo-2,1,3-thiadiazole-4-sulfonyl chloride, 2??,1,3-benzothiadiazole-4-sulfonyl chloride, benzo-2,1,3-thiadiazole-4-sulphonyl chloride
PubChem CID 2735459
IUPAC Name 2,1,3-benzothiadiazole-4-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=CC2=NSN=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 234.69
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzothiadiazole-4-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.