missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazole-4-carbaldehyde, 95%, Thermo Scientific™

Catalog No. p-2696547
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC09104CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC09104CB Supplier Thermo Scientific Chemicals Supplier No. CC09104CB

Chemical Identifiers

CAS 5170-68-3
Molecular Formula C7H4N2OS
Molecular Weight (g/mol) 164.182
MDL Number MFCD02681902
InChI Key ANVJARPTPIVPRC-UHFFFAOYSA-N
Synonym benzo c 1,2,5 thiadiazole-4-carbaldehyde, 2,1,3-benzothiadiazole-4-carboxaldehyde, tos-bb-1233, 2,1,3-benzothiadiazol-4-aldehyde, 2,1,3-benzothiadiazole-4-aldehyde, 2,1,3-benzothiadiazol-4-carboxaldehyde, benzo c 1,2,5-thiadiazole-4-carbaldehyde, 2,1,3-benzothiadiazole-4-carbaldehyde
PubChem CID 2776283
IUPAC Name 2,1,3-benzothiadiazole-4-carbaldehyde
SMILES C1=CC2=NSN=C2C(=C1)C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 164.19
Percent Purity 95%
Chemical Name or Material 2, 1, 3-Benzothiadiazole-4-carbaldehyde