missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazol-5-yl isothiocyanate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 337508-62-0
Molecular Formula C7H3N3S2
Molecular Weight (g/mol) 193.242
MDL Number MFCD02681895
InChI Key HHDWFDQYUFKNCO-UHFFFAOYSA-N
Synonym 2,1,3-benzothiadiazol-5-yl isothiocyanate, 2,1,3-benzothiadiazol-5-ylisothiocyanate, 2,1,3-benzothiadiazole, 5-isothiocyanato, 2,1,3-benzothiadiazole,5-isothiocyanato, benzo c 1,2,5-thiadiazol-5-isothiocyanate, 5-isothiocyanatobenzo c 1.2.5 thiadiazole
PubChem CID 2776292
IUPAC Name 5-isothiocyanato-2,1,3-benzothiadiazole
SMILES C1=CC2=NSN=C2C=C1N=C=S
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC09207CB
SDS View Documents
Thermo Scientific Chemicals
CC09207CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

337508-62-0
193.242
HHDWFDQYUFKNCO-UHFFFAOYSA-N
2776292
C1=CC2=NSN=C2C=C1N=C=S
C7H3N3S2
MFCD02681895
2,1,3-benzothiadiazol-5-yl isothiocyanate, 2,1,3-benzothiadiazol-5-ylisothiocyanate, 2,1,3-benzothiadiazole, 5-isothiocyanato, 2,1,3-benzothiadiazole,5-isothiocyanato, benzo c 1,2,5-thiadiazol-5-isothiocyanate, 5-isothiocyanatobenzo c 1.2.5 thiadiazole
5-isothiocyanato-2,1,3-benzothiadiazole

Specifications

337508-62-0
100.0
Amber glass bottle
MFCD02681895
2,1,3-benzothiadiazol-5-yl isothiocyanate, 2,1,3-benzothiadiazol-5-ylisothiocyanate, 2,1,3-benzothiadiazole, 5-isothiocyanato, 2,1,3-benzothiadiazole,5-isothiocyanato, benzo c 1,2,5-thiadiazol-5-isothiocyanate, 5-isothiocyanatobenzo c 1.2.5 thiadiazole
C1=CC2=NSN=C2C=C1N=C=S
193.242
193.25
2, 1, 3-Benzothiadiazol-5-yl isothiocyanate
96.5
97%
C7H3N3S2
250mg
HHDWFDQYUFKNCO-UHFFFAOYSA-N
5-isothiocyanato-2,1,3-benzothiadiazole
2776292
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'