missing translation for 'onlineSavingsMsg'
Learn More

2-(1,3-Benzodioxol-5-yl)ethanol, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC50809DA

Quantity 1g

 View more versions of this product

Catalog No. CC50809DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

6006-82-2
166.176
JADSGOFBFPTCHG-UHFFFAOYSA-N
2759846
C1OC2=C(O1)C=C(C=C2)CCO
C9H10O3
MFCD00666033
2-benzo d 1,3 dioxol-5-yl ethanol, 2-2h-1,3-benzodioxol-5-yl ethan-1-ol, 3,4-methylenedioxy phenethyl alcohol, 2-1,3-benzodioxol-5-yl ethanol, 1,3-benzodioxole-5-ethanol, homopiperonyl alcohol, 3,4-methylenedioxyphenethyl alcohol, 2-1,3-benzodioxol-5-yl-ethanol, 2-2h-1,3-benzodioxol-5-yl ethanol, benzodioxolylethanol
2-(1,3-benzodioxol-5-yl)ethanol

Specifications

2-(1, 3-Benzodioxol-5-yl)ethanol
96.5
97%
C9H10O3
MFCD00666033
JADSGOFBFPTCHG-UHFFFAOYSA-N
2-(1,3-benzodioxol-5-yl)ethanol
2759846
97%
6006-82-2
100.0
Amber Glass Bottle
1g
2-benzo d 1,3 dioxol-5-yl ethanol, 2-2h-1,3-benzodioxol-5-yl ethan-1-ol, 3,4-methylenedioxy phenethyl alcohol, 2-1,3-benzodioxol-5-yl ethanol, 1,3-benzodioxole-5-ethanol, homopiperonyl alcohol, 3,4-methylenedioxyphenethyl alcohol, 2-1,3-benzodioxol-5-yl-ethanol, 2-2h-1,3-benzodioxol-5-yl ethanol, benzodioxolylethanol
C1OC2=C(O1)C=C(C=C2)CCO
166.176
166.18
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents