missing translation for 'onlineSavingsMsg'
Learn More

2-(1,3-Benzodioxol-5-yl)ethanol, ≥97%, Thermo Scientific™

Catalog No. CC50809CB
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC50809CB 250mg
CC50809DA 1g
CC50809DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC50809CB Supplier Thermo Scientific Chemicals Supplier No. CC50809CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6006-82-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00666033
InChI Key JADSGOFBFPTCHG-UHFFFAOYSA-N
Synonym 2-benzo d 1,3 dioxol-5-yl ethanol, 2-2h-1,3-benzodioxol-5-yl ethan-1-ol, 3,4-methylenedioxy phenethyl alcohol, 2-1,3-benzodioxol-5-yl ethanol, 1,3-benzodioxole-5-ethanol, homopiperonyl alcohol, 3,4-methylenedioxyphenethyl alcohol, 2-1,3-benzodioxol-5-yl-ethanol, 2-2h-1,3-benzodioxol-5-yl ethanol, benzodioxolylethanol
PubChem CID 2759846
IUPAC Name 2-(1,3-benzodioxol-5-yl)ethanol
SMILES C1OC2=C(O1)C=C(C=C2)CCO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material 2-(1, 3-Benzodioxol-5-yl)ethanol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.