missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(-)-alpha-Pinene, 98%

CAS: 7785-26-4 | C10H16 | 136.24 g/mol

Supplier:  Thermo Scientific Chemicals 131272500

Quantity 250 mL
Packaging Glass bottle

 View more versions of this product

Catalog No. AC131272500


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7785-26-4
136.24
--alpha-pinene, 1s,5s-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1s---alpha-pinene, unii-tzr3gm95pr, dl-alpha-pinene, alpha-pinene,-, 1s,5s-alpha-pinene, tzr3gm95pr, 1s,5s-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1s,5s
CHEBI:28660
CC1=CCC2CC1C2(C)C
C10H16
GRWFGVWFFZKLTI-IUCAKERBSA-N
440968
(1S,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene

Specifications

(1S)-(-)-α-Pinene
-64.0°C
0.8595g/mL
33°C
97.5% min. (GC)
C10H16
250 mL
14,7445
- 41.50 (20.00°C c=neat)
--alpha-pinene, 1s,5s-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1s---alpha-pinene, unii-tzr3gm95pr, dl-alpha-pinene, alpha-pinene,-, 1s,5s-alpha-pinene, tzr3gm95pr, 1s,5s-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1s,5s
GRWFGVWFFZKLTI-IUCAKERBSA-N
(1S,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene
440968
136.24
Liquid
7785-26-4
Colorless
155.0°C to 156.0°C
Authentic
Glass bottle
1.4650 to 1.4670
05,146
0.8595
− 41.50
Solubility in water: insoluble. Other solubilities: soluble in chloroform,alcohol and ether
CC1=CCC2CC1C2(C)C
136.24
CHEBI:28660
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting

GHS P Statement
Wear protective gloves/protective clothing.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
If skin irritation or rash occurs: Get medical advice/attention.
IF ON SKIN (or hair): Take off immediately all contam

GHS Signal Word: Danger

EINECSNumber : 232-077-3

SDS
Documents

Documents

Promotions

Promotions