missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(-)-Camphorsulfonylimine, 98+%

Catalog No. AAB2082314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2082314 25 g
AAB2082303 1 g
AAB2082306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2082314 Supplier Thermo Scientific Chemicals Supplier No. B2082314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 60886-80-8 | C10H15NO2S | 213.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60886-80-8
Molecular Formula C10H15NO2S
Molecular Weight (g/mol) 213.30
MDL Number MFCD00013315,MFCD00064576
InChI Key ZAHOEBNYVSWBBW-UHFFFAOYNA-N
Synonym 1s,7s-10,10-dimethyl-3-thia-4-azatricyclo 5.2.1.0<1,5> dec-4-ene-3,3-dione, 1s,7s-10,10-dimethyl-3??-thia-4-azatricyclo 5.2.1.0 1 ,? dec-4-ene-3,3-dione
PubChem CID 45925640
IUPAC Name 10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]dec-4-ene-3,3-dione
SMILES CC1(C)C2CCC11CS(=O)(=O)N=C1C2

Specifications

Melting Point 226°C to 228°C
Quantity 25 g
Beilstein 85296
Formula Weight 213.3
Percent Purity ≥98%
Chemical Name or Material (1S)-(-)-Camphorsulfonylimine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only