missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(+)-Camphor-10-sulfonic acid, 98+%(dry wt.), water <2%

CAS: 3144-16-9 | C10H15O4S | 231.29 g/mol

Supplier:  Thermo Scientific Chemicals A1617922

Quantity 100 g

 View more versions of this product

Catalog No. AAA1617922


Only null left

Description

Description

(1S)-(+)-Camphor-10-sulfonic acid is used as stabilizer. (+)-(1S)-camphor-10-sulfonic acid being a very effective resolving agent. . Chiral polyaniline(PANI) nanofibers were synthesized via facilely potentiostatic electropolymerization method without template in the presence of(1S)-(+)camphor-10-sulfonic acid(D-CSA) or(1R)-(-)camphor-10-sulfonic acid(L-CSA) as the dopant. Synthesis of QUATs derived from (1S)-(+)camphor-10-sulfonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S)-(+)-Camphor-10-sulfonic acid is used as stabilizer. (+)-(1S)-camphor-10-sulfonic acid being a very effective resolving agent. . Chiral polyaniline(PANI) nanofibers were synthesized via facilely potentiostatic electropolymerization method without template in the presence of(1S)-(+)camphor-10-sulfonic acid(D-CSA) or(1R)-(-)camphor-10-sulfonic acid(L-CSA) as the dopant. Synthesis of QUATs derived from (1S)-(+)camphor-10-sulfonic acid.

Solubility
Exhibit moderate solubility in HCCl.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases. Keep under dry inert gas.
Specifications

Chemical Identifiers

3144-16-9
231.29
MIOPJNTWMNEORI-XVKPBYJWSA-M
65617
[(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
C10H15O4S
MFCD00064157,MFCD00074827
1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid
CHEBI:55403
CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2

Specifications

(1S)-(+)-Camphor-10-sulfonic acid
196°C to 200°C (decomposition)
C10H15O4S
MFCD00064157,MFCD00074827
2809675
14,1734
Exhibit moderate solubility in HCCl.
CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2
[(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
65617
232.3
3144-16-9
(dry wt.), water <2%
100 g
UN3261
Hygroscopic
1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid
MIOPJNTWMNEORI-XVKPBYJWSA-M
+20° (c=2 in Water)
231.29
CHEBI:55403
≥98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 221-554-1

RTECSNumber : ED1530000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions