missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(+)-Camphor-10-sulfonic acid, 98+%(dry wt.), water <2%

Catalog No. AAA1617914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1617914 25 g
AAA1617922 100 g
AAA1617936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1617914 Supplier Thermo Scientific Chemicals Supplier No. A1617914

CAS: 3144-16-9 | C10H15O4S | 231.29 g/mol

(1S)-(+)-Camphor-10-sulfonic acid is used as stabilizer. (+)-(1S)-camphor-10-sulfonic acid being a very effective resolving agent. . Chiral polyaniline(PANI) nanofibers were synthesized via facilely potentiostatic electropolymerization method without template in the presence of(1S)-(+)camphor-10-sulfonic acid(D-CSA) or(1R)-(-)camphor-10-sulfonic acid(L-CSA) as the dopant. Synthesis of QUATs derived from (1S)-(+)camphor-10-sulfonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S)-(+)-Camphor-10-sulfonic acid is used as stabilizer. (+)-(1S)-camphor-10-sulfonic acid being a very effective resolving agent. . Chiral polyaniline(PANI) nanofibers were synthesized via facilely potentiostatic electropolymerization method without template in the presence of(1S)-(+)camphor-10-sulfonic acid(D-CSA) or(1R)-(-)camphor-10-sulfonic acid(L-CSA) as the dopant. Synthesis of QUATs derived from (1S)-(+)camphor-10-sulfonic acid.

Solubility
Exhibit moderate solubility in HCCl.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases. Keep under dry inert gas.

Chemical Identifiers

CAS 3144-16-9
Molecular Formula C10H15O4S
Molecular Weight (g/mol) 231.29
MDL Number MFCD00064157,MFCD00074827
InChI Key MIOPJNTWMNEORI-XVKPBYJWSA-M
Synonym 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid
PubChem CID 65617
ChEBI CHEBI:55403
IUPAC Name [(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
SMILES CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2

Specifications

Melting Point 196°C to 200°C (decomposition)
Quantity 25 g
UN Number UN3261
Beilstein 2809675
Sensitivity Hygroscopic
Merck Index 14,1734
Solubility Information Exhibit moderate solubility in HCCl.
Optical Rotation +20° (c=2 in Water)
Formula Weight 232.3
Percent Purity ≥98%
Assay (dry wt.), water <2%
Chemical Name or Material (1S)-(+)-Camphor-10-sulfonic acid
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 221-554-1
RTECSNumber ED1530000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only