missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(-)-alpha-Pinene 97.0+%, TCI America™

Catalog No. P044025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P044025ML 25 mL
P0440500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P044025ML Supplier TCI America Supplier No. P044025ML

Chemical Identifiers

CAS 7785-26-4
Molecular Formula C10H16
Molecular Weight (g/mol) 136.238
MDL Number MFCD00064145
InChI Key GRWFGVWFFZKLTI-IUCAKERBSA-N
Synonym --alpha-pinene, 1s,5s-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1s---alpha-pinene, unii-tzr3gm95pr, dl-alpha-pinene, alpha-pinene,-, 1s,5s-alpha-pinene, tzr3gm95pr, 1s,5s-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1s,5s
PubChem CID 440968
ChEBI CHEBI:28660
IUPAC Name (1S,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene
SMILES CC1=CCC2CC1C2(C)C

Specifications

Melting Point -64°C
Color Yellow
Boiling Point 155°C
Quantity 25 mL
UN Number 2368
Formula Weight 136.24
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (1S)-(-)-alpha-Pinene
EINECSNumber (4)-0593&(8)-0497
RTECSNumber DT7000000
TSCA Yes