missing translation for 'onlineSavingsMsg'
Learn More

(1S,4S,5S)-4,7,7-Trimethyl-6-thiabicyclo[3.2.1]octane 98.0+%, TCI America™

Catalog No. T25791G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25791G 1 g
T25795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25791G Supplier TCI America Supplier No. T25791G

Chemical Identifiers

CAS 1208985-45-8
Molecular Formula C10H18S
Molecular Weight (g/mol) 170.31
MDL Number MFCD18207721
InChI Key FAXNZPOZWCWYBD-UHFFFAOYNA-N
PubChem CID 56924133
IUPAC Name 4,7,7-trimethyl-6-thiabicyclo[3.2.1]octane
SMILES CC1CCC2CC1SC2(C)C

Specifications

Color Yellow
Quantity 1 g
Formula Weight 170.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (1S,4S,5S)-4,7,7-Trimethyl-6-thiabicyclo[3.2.1]octane