missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,4R)-N-BOC-1-Aminocyclopent-2-ene-4-carboxylic acid, 95%, 98% ee, Thermo Scientific™
Chemical Identifiers
| CAS | 151907-80-1 |
|---|---|
| Molecular Formula | C11H17NO4 |
| Molecular Weight (g/mol) | 227.26 |
| MDL Number | MFCD00211288 |
| InChI Key | WOUNTSATDZJBLP-UHFFFAOYNA-N |
| Synonym | 1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonylamino cyclopent-2-enecarboxylic acid, 1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxylic acid, +-1r,4s-n-boc-4-aminocyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-ene-1-carboxylic acid, +-1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxy, cis-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxy carbonyl amino cyclopent-2-ene-1-carboxylic acid, 1r,4s-rel-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid |
| PubChem CID | 7019281 |
| IUPAC Name | (1R,4S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopent-2-ene-1-carboxylic acid |
| SMILES | CC(C)(C)OC(=O)NC1CC(C=C1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC361820010
|
N/A | 1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 151907-80-1 | |
| 227.26 | |
| WOUNTSATDZJBLP-UHFFFAOYNA-N | |
| 7019281 | |
| CC(C)(C)OC(=O)NC1CC(C=C1)C(O)=O |
| C11H17NO4 | |
| MFCD00211288 | |
| 1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonylamino cyclopent-2-enecarboxylic acid, 1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxylic acid, +-1r,4s-n-boc-4-aminocyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-ene-1-carboxylic acid, +-1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxy, cis-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxy carbonyl amino cyclopent-2-ene-1-carboxylic acid, 1r,4s-rel-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid | |
| (1R,4S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopent-2-ene-1-carboxylic acid |
Specifications
| 151907-80-1 | |
| 100.0 | |
| Authentic | |
| C11H17NO4 | |
| 1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonylamino cyclopent-2-enecarboxylic acid, 1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxylic acid, +-1r,4s-n-boc-4-aminocyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-ene-1-carboxylic acid, +-1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxy, cis-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxy carbonyl amino cyclopent-2-ene-1-carboxylic acid, 1r,4s-rel-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid | |
| WOUNTSATDZJBLP-UHFFFAOYNA-N | |
| (1R,4S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopent-2-ene-1-carboxylic acid | |
| 7019281 | |
| 95%,98% ee |
| 94.0 | |
| 97.5% min. (HPLC) | |
| 94% min. (HPLC) | |
| MFCD00211288 | |
| Solubility in water: soluble | |
| CC(C)(C)OC(=O)NC1CC(C=C1)C(O)=O | |
| 227.26 | |
| 227.26 | |
| (1S, 4R)-N-BOC-1-Aminocyclopent-2-ene-4-carboxylic acid, 98% ee |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Warning
RUO – Research Use Only