missing translation for 'onlineSavingsMsg'
Learn More

(1S,3R)-cis-3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid, 97%

Catalog No. AAH52747MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52747MD 250 mg
AAH5274703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52747MD Supplier Thermo Scientific Chemicals Supplier No. H52747MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96443-42-4 | C8H12O4 | 172.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96443-42-4
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD04972280
InChI Key FVUHGTQDOMGZOT-NTSWFWBYSA-N
Synonym 1s,3r-3-methoxycarbonyl cyclopentanecarboxylic acid, 1s,3r-3-methoxycarbonyl cyclopentane-1-carboxylic acid, 1r,3s-1,3-cyclopentanedicarboxylic acid 1-methyl ester, 1s,3r-cis-3-carbomethoxy cyclopentane-1-carboxylic acid, cis-3-methoxycarbonyl cyclopentancarboxylic acid, 1s,3r-cis-3-carbomethoxy cyclopentane-1-carboxyl, 1,3-cyclopentanedicarboxylicacid,1-methyl ester, 1r,3s, 1r-cis-1,3-cyclopentanedicarboxylic acid monomethyl ester, 1,3-cyclopentanedicarboxylic acid, 1-methyl ester, 1r,3s
PubChem CID 10888375
IUPAC Name (1S,3R)-3-methoxycarbonylcyclopentane-1-carboxylic acid
SMILES COC(=O)C1CCC(C1)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 172.18
Percent Purity 97%
Chemical Name or Material (1S,3R)-cis-3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only