missing translation for 'onlineSavingsMsg'
Learn More

(1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic Acid 95.0+%, TCI America™

Catalog No. B4397200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4397200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4397200MG Supplier TCI America Supplier No. B4397200MG
Only null left

Chemical Identifiers

CAS 261165-05-3
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD01320857
InChI Key RNJQBGXOSAQQDG-JGVFFNPUSA-N
Synonym 1s,3r-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, +-1s,3r-n-boc-3-aminocyclopentanecarboxylic acid, 1r,3s-n-boc-1-aminocyclopentane-3-carboxylic acid, 1s,3r-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, cis-3-boc-amino cyclopentane-1-carboxylic acid, 1s,3r-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1s,3r-3-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid
PubChem CID 1512529
IUPAC Name (1S,3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCC(C1)C(=O)O

Specifications

Melting Point 111°C
Color White-Yellow
Quantity 200 mg
Formula Weight 229.28
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic Acid