missing translation for 'onlineSavingsMsg'
Learn More

(1S,3R)-3-Aminocyclopentanecarboxylic acid, 95%, >90% ee

Catalog No. AC362280010
Change view
Click to view available options
Quantity:
1 g
5g, 5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC362280010 1 g Glass Bottle
AC362280050 5g, 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC362280010 Supplier Thermo Scientific Chemicals Supplier No. 362280010
Only null left

CAS: 71830-07-4 | C6H11NO2 | 129.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71830-07-4
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00211290
InChI Key MLLSSTJTARJLHK-CRCLSJGQSA-N
Synonym 1s,3r-3-aminocyclopentanecarboxylic acid, 1s,3r-3-aminocyclopentane-1-carboxylic acid, +-1s,3r-3-aminocyclopentanecarboxylic acid, 1s,3r-3-amino-cyclopentanecarboxylic acid, cis-3-aminocyclopentanecarboxylic acid, 1s,3r-+-3-aminocyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-amino-, 1s,3r, pubchem18730, +-cacp, cis-3-aminocyclopentane-1-carboxylic acid
PubChem CID 1502034
IUPAC Name (1S,3R)-3-aminocyclopentane-1-carboxylic acid
SMILES C1CC(CC1C(=O)O)N

Specifications

Melting Point 192.0°C
CAS Min % 94.0
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 129.16
Percent Purity 95%,>90% ee
Physical Form Crystalline Powder
Chemical Name or Material (1S,3R)-3-Aminocyclopentanecarboxylic acid, >90% ee
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

RUO – Research Use Only