missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,3R)-3-Aminocyclopentanecarboxylic acid, 95%, >90% ee
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 192.0°C |
| CAS Min % | 94.0 |
| CAS Max % | 100.0 |
| Color | White to Beige |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 95% |
| Packaging | Glass Bottle |
| Quantity | 1 g |
| Formula Weight | 129.16 |
| Percent Purity | 95%,>90% ee |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Warning |
| missing translation for 'healthHazard2' | GHS H Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation. |
| missing translation for 'healthHazard3' | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor |
RUO – Research Use Only