missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane, 98%

CAS: 87583-89-9 | C8H18N2 | 142.246 g/mol

Supplier:  Thermo Scientific Chemicals H6439606

 View more versions of this product

Catalog No. AAH6439606

Only null left
Explore available promotions

Description

Description

It is employed in the synthesis of 6-substituted 1-oxoindanoyl isoleucine conjugates and modeling studies with the coi1-jaz co-receptor complex of lima bean.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of 6-substituted 1-oxoindanoyl isoleucine conjugates and modeling studies with the coi1-jaz co-receptor complex of lima bean.

Solubility
Slightly soluble in water.

Notes
Store away from acids, air, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Store under inert gas. Keep it refrigerated.
Specifications

Specifications

(1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane
39°C to 44°C
0.902 g/mL
C8H18N2
MFCD00671528
UN3259
1s,2s-n1,n2-dimethylcyclohexane-1,2-diamine, 1s,2s-n,n'-dimethyl-1,2-cyclohexanediamine, 1s,2s-+-n,n'-dimethylcyclohexane-1,2-diamine, trans-1s,2s-n,n'-dimethylcyclohexane-1,2-diamine, 1s,2s-+-1,2-bis methylamino cyclohexane, 1s,2s-n,n'-bismethyl-1,2-cyclohexane-diamine, 1s,2s-1-n,2-n-dimethylcyclohexane-1,2-diamine, 1s,2s-n,n'-dimethyl-cyclohexane-1,2-diamine, n,n'-dimethyl-1s,2s-1,2-cyclohexanediamine, trans-1s,2s-n,n-dimethyl-cyclohexane-1,2-diamine
JRHPOFJADXHYBR-YUMQZZPRSA-N
(1S,2S)-1-N,2-N-dimethylcyclohexane-1,2-diamine
13822957
98%
87583-89-9
Brown
74°C (165°F)
1.472
5 g
4304824
Slightly soluble in water.
CNC1CCCCC1NC
142.246
142.25
Solid

Chemical Identifiers

87583-89-9
142.246
JRHPOFJADXHYBR-YUMQZZPRSA-N
13822957
CNC1CCCCC1NC
C8H18N2
MFCD00671528
1s,2s-n1,n2-dimethylcyclohexane-1,2-diamine, 1s,2s-n,n'-dimethyl-1,2-cyclohexanediamine, 1s,2s-+-n,n'-dimethylcyclohexane-1,2-diamine, trans-1s,2s-n,n'-dimethylcyclohexane-1,2-diamine, 1s,2s-+-1,2-bis methylamino cyclohexane, 1s,2s-n,n'-bismethyl-1,2-cyclohexane-diamine, 1s,2s-1-n,2-n-dimethylcyclohexane-1,2-diamine, 1s,2s-n,n'-dimethyl-cyclohexane-1,2-diamine, n,n'-dimethyl-1s,2s-1,2-cyclohexanediamine, trans-1s,2s-n,n-dimethyl-cyclohexane-1,2-diamine
(1S,2S)-1-N,2-N-dimethylcyclohexane-1,2-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only