missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane, 98%

Catalog No. AAH6439614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6439614 25 g
AAH6439603 1 g
AAH6439606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6439614 Supplier Thermo Scientific Chemicals Supplier No. H6439614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 87583-89-9 | C8H18N2 | 142.246 g/mol

It is employed in the synthesis of 6-substituted 1-oxoindanoyl isoleucine conjugates and modeling studies with the coi1-jaz co-receptor complex of lima bean.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of 6-substituted 1-oxoindanoyl isoleucine conjugates and modeling studies with the coi1-jaz co-receptor complex of lima bean.

Solubility
Slightly soluble in water.

Notes
Store away from acids, air, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Store under inert gas. Keep it refrigerated.

Chemical Identifiers

CAS 87583-89-9
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD00671528
InChI Key JRHPOFJADXHYBR-YUMQZZPRSA-N
Synonym 1s,2s-n1,n2-dimethylcyclohexane-1,2-diamine, 1s,2s-n,n'-dimethyl-1,2-cyclohexanediamine, 1s,2s-+-n,n'-dimethylcyclohexane-1,2-diamine, trans-1s,2s-n,n'-dimethylcyclohexane-1,2-diamine, 1s,2s-+-1,2-bis methylamino cyclohexane, 1s,2s-n,n'-bismethyl-1,2-cyclohexane-diamine, 1s,2s-1-n,2-n-dimethylcyclohexane-1,2-diamine, 1s,2s-n,n'-dimethyl-cyclohexane-1,2-diamine, n,n'-dimethyl-1s,2s-1,2-cyclohexanediamine, trans-1s,2s-n,n-dimethyl-cyclohexane-1,2-diamine
PubChem CID 13822957
IUPAC Name (1S,2S)-1-N,2-N-dimethylcyclohexane-1,2-diamine
SMILES CNC1CCCCC1NC

Specifications

Melting Point 39°C to 44°C
Color Brown
Density 0.902 g/mL
Flash Point 74°C (165°F)
Refractive Index 1.472
Quantity 25 g
UN Number UN3259
Beilstein 4304824
Solubility Information Slightly soluble in water.
Formula Weight 142.25
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only