missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(+)-N,N'-Dimethylcyclohexane-1,2-diamine 98.0+%, TCI America™

Catalog No. D2460100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2460100MG 100 mg
D24601G 1 g
D24605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D2460100MG Supplier TCI America Supplier No. D2460100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87583-89-9
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD00671528
InChI Key JRHPOFJADXHYBR-YUMQZZPRSA-N
Synonym 1s,2s-n1,n2-dimethylcyclohexane-1,2-diamine, 1s,2s-n,n'-dimethyl-1,2-cyclohexanediamine, 1s,2s-+-n,n'-dimethylcyclohexane-1,2-diamine, trans-1s,2s-n,n'-dimethylcyclohexane-1,2-diamine, 1s,2s-+-1,2-bis methylamino cyclohexane, 1s,2s-n,n'-bismethyl-1,2-cyclohexane-diamine, 1s,2s-1-n,2-n-dimethylcyclohexane-1,2-diamine, 1s,2s-n,n'-dimethyl-cyclohexane-1,2-diamine, n,n'-dimethyl-1s,2s-1,2-cyclohexanediamine, trans-1s,2s-n,n-dimethyl-cyclohexane-1,2-diamine
PubChem CID 13822957
IUPAC Name (1S,2S)-1-N,2-N-dimethylcyclohexane-1,2-diamine
SMILES CNC1CCCCC1NC

Specifications

Melting Point 47°C
Color White-Yellow
Quantity 100 mg
UN Number 3259
Formula Weight 142.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (1S,2S)-(+)-N,N′-Dimethylcyclohexane-1,2-diamine
TSCA No
Recommended Storage Refrigerator