missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-N,N'-Dimethyl-N,N'-bis(3,3-dimethylbutyl)cyclohexane-1,2-diamine 94.0+%, TCI America™

Catalog No. D38091G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D38091G Supplier TCI America Supplier No. D38091G
Only null left

Chemical Identifiers

CAS 767291-67-8
Molecular Formula C20H42N2
Molecular Weight (g/mol) 310.57
InChI Key VGCWVKVNKNXOGZ-ROUUACIJSA-N
PubChem CID 11392766
IUPAC Name (1S,2S)-1-N,2-N-bis(3,3-dimethylbutyl)-1-N,2-N-dimethylcyclohexane-1,2-diamine
SMILES CC(C)(C)CCN(C)C1CCCCC1N(C)CCC(C)(C)C

Specifications

Color Yellow
Boiling Point 145°C
Quantity 1 g
Formula Weight 310.57
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material (1S,2S)-N,N′-Dimethyl-N,N′-bis(3,3-dimethylbutyl)cyclohexane-1,2-diamine