missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,2S,5S)-(-)-2-Hydroxy-3-pinanone 98.0+%, TCI America™
$152.86 - $600.22
Chemical Identifiers
| CAS | 1845-25-6 |
|---|---|
| Molecular Formula | C10H16O2 |
| Molecular Weight (g/mol) | 168.236 |
| MDL Number | MFCD00040556 |
| InChI Key | VZRRCQOUNSHSGB-BYULHYEWSA-N |
| Synonym | (1S,2S,5S)-(-)-2-Hydroxy-2,6,6-trimethylbicyclo[3.1.1]heptan-3-one |
| PubChem CID | 11126668 |
| IUPAC Name | (1S,4S,5S)-4-hydroxy-4,6,6-trimethylbicyclo[3.1.1]heptan-3-one |
| SMILES | CC1(C2CC1C(C(=O)C2)(C)O)C |
Chemical Identifiers
| 1845-25-6 | |
| 168.236 | |
| VZRRCQOUNSHSGB-BYULHYEWSA-N | |
| 11126668 | |
| CC1(C2CC1C(C(=O)C2)(C)O)C |
| C10H16O2 | |
| MFCD00040556 | |
| (1S,2S,5S)-(-)-2-Hydroxy-2,6,6-trimethylbicyclo[3.1.1]heptan-3-one | |
| (1S,4S,5S)-4-hydroxy-4,6,6-trimethylbicyclo[3.1.1]heptan-3-one |
Specifications
| 1845-25-6 | |
| C10H16O2 | |
| 1 g | |
| VZRRCQOUNSHSGB-BYULHYEWSA-N | |
| (1S,4S,5S)-4-hydroxy-4,6,6-trimethylbicyclo[3.1.1]heptan-3-one | |
| 11126668 | |
| ≥98.0% (GC) | |
| (1S,2S,5S)-(-)-2-Hydroxy-3-pinanone |
| White-Yellow | |
| MFCD00040556 | |
| (1S,2S,5S)-(-)-2-Hydroxy-2,6,6-trimethylbicyclo[3.1.1]heptan-3-one | |
| CC1(C2CC1C(C(=O)C2)(C)O)C | |
| 168.236 | |
| 168.24 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No