missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S,5S)-(-)-2-Hydroxy-3-pinanone 98.0+%, TCI America™

Catalog No. H08631G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08631G 1 g
H08635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08631G Supplier TCI America Supplier No. H08631G
Only null left

Chemical Identifiers

CAS 1845-25-6
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.236
MDL Number MFCD00040556
InChI Key VZRRCQOUNSHSGB-BYULHYEWSA-N
Synonym (1S,2S,5S)-(-)-2-Hydroxy-2,6,6-trimethylbicyclo[3.1.1]heptan-3-one
PubChem CID 11126668
IUPAC Name (1S,4S,5S)-4-hydroxy-4,6,6-trimethylbicyclo[3.1.1]heptan-3-one
SMILES CC1(C2CC1C(C(=O)C2)(C)O)C

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 168.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (1S,2S,5S)-(-)-2-Hydroxy-3-pinanone