missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,2S,4S,5S)-2,5-Bis(3,5-di-tert-butyl-2-hydroxybenzylideneamino)bicyclo[2.2.1]heptane 98.0+%, TCI America™
Supplier: TCI America B2653100MG
Specifications
| (1S,2S,4S,5S)-2,5-Bis(3,5-di-tert-butyl-2-hydroxybenzylideneamino)bicyclo[2.2.1]heptane | |
| 254°C | |
| C37H54N2O2 | |
| 100 mg | |
| SMGJFOOBGSAVJS-UHFFFAOYNA-N | |
| 2,4-di-tert-butyl-6-{[(5-{[(3,5-di-tert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}bicyclo[2.2.1]heptan-2-yl)amino]methylidene}cyclohexa-2,4-dien-1-one | |
| 90473853 | |
| ≥98.0% (HPLC,N) |
| 539834-16-7 | |
| Yellow | |
| MFCD08276289,MFCD08276290 | |
| 2,2′C-[(1S,2S,4S,5S)-Bicyclo[2.2.1]heptane-2,5-diylbis(nitrilomethyl)]bis(4,6-di-tert-butylphenol) | |
| CC(C)(C)C1=CC(=CNC2CC3CC2CC3NC=C2C=C(C=C(C2=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C | |
| 558.85 | |
| 558.85 | |
| Crystalline Powder |
Chemical Identifiers
| 539834-16-7 | |
| 558.85 | |
| SMGJFOOBGSAVJS-UHFFFAOYNA-N | |
| 90473853 | |
| CC(C)(C)C1=CC(=CNC2CC3CC2CC3NC=C2C=C(C=C(C2=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C |
| C37H54N2O2 | |
| MFCD08276289,MFCD08276290 | |
| 2,2′C-[(1S,2S,4S,5S)-Bicyclo[2.2.1]heptane-2,5-diylbis(nitrilomethyl)]bis(4,6-di-tert-butylphenol) | |
| 2,4-di-tert-butyl-6-{[(5-{[(3,5-di-tert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}bicyclo[2.2.1]heptan-2-yl)amino]methylidene}cyclohexa-2,4-dien-1-one |
Safety and Handling
TSCA : No