missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,2S,3R,5S)-(+)-2,3-Pinanediol 98.0+%, TCI America™
$104.43 - $332.26
Chemical Identifiers
| CAS | 18680-27-8 |
|---|---|
| Molecular Formula | C10H18O2 |
| Molecular Weight (g/mol) | 170.252 |
| MDL Number | MFCD00077851 |
| InChI Key | MOILFCKRQFQVFS-OORONAJNSA-N |
| Synonym | 1s,2s,3r,5s-+-2,3-pinanediol, 1s,2s,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, 1s,2s,3r,5s-+-pinanediol, +-pinanediol, pinanediol, +, unii-y6zcv4avra, +-2-hydroxyisopinocampheol, y6zcv4avra, 1s,2s,3r,5s-2,3-pinanediol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1s,2s,3r,5s |
| PubChem CID | 10219606 |
| IUPAC Name | (1S,3R,4S,5S)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol |
| SMILES | CC1(C2CC1C(C(C2)O)(C)O)C |
Chemical Identifiers
| 18680-27-8 | |
| 170.252 | |
| MOILFCKRQFQVFS-OORONAJNSA-N | |
| 10219606 | |
| CC1(C2CC1C(C(C2)O)(C)O)C |
| C10H18O2 | |
| MFCD00077851 | |
| 1s,2s,3r,5s-+-2,3-pinanediol, 1s,2s,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, 1s,2s,3r,5s-+-pinanediol, +-pinanediol, pinanediol, +, unii-y6zcv4avra, +-2-hydroxyisopinocampheol, y6zcv4avra, 1s,2s,3r,5s-2,3-pinanediol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1s,2s,3r,5s | |
| (1S,3R,4S,5S)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol |
Specifications
| 18680-27-8 | |
| C10H18O2 | |
| 1s,2s,3r,5s-+-2,3-pinanediol, 1s,2s,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, 1s,2s,3r,5s-+-pinanediol, +-pinanediol, pinanediol, +, unii-y6zcv4avra, +-2-hydroxyisopinocampheol, y6zcv4avra, 1s,2s,3r,5s-2,3-pinanediol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1s,2s,3r,5s | |
| CC1(C2CC1C(C(C2)O)(C)O)C | |
| 170.252 | |
| 170.25 | |
| Crystalline Powder |
| 57°C | |
| MFCD00077851 | |
| MOILFCKRQFQVFS-OORONAJNSA-N | |
| (1S,3R,4S,5S)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol | |
| 10219606 | |
| ≥98.0% (GC) | |
| (1S,2S,3R,5S)-(+)-2,3-Pinanediol |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator