missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S,3R,5S)-(+)-2,3-Pinanediol, 98%

Catalog No. AC279550025
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC279550025 2.5 g Glass bottle
AC279550100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC279550025 Supplier Thermo Scientific Chemicals Supplier No. 279550025
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 18680-27-8 | C10H18O2 | 170.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18680-27-8
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00077851
InChI Key MOILFCKRQFQVFS-OORONAJNSA-N
Synonym 1s,2s,3r,5s-+-2,3-pinanediol, 1s,2s,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, 1s,2s,3r,5s-+-pinanediol, +-pinanediol, pinanediol, +, unii-y6zcv4avra, +-2-hydroxyisopinocampheol, y6zcv4avra, 1s,2s,3r,5s-2,3-pinanediol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1s,2s,3r,5s
PubChem CID 10219606
IUPAC Name (1S,3R,4S,5S)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol
SMILES CC1(C2CC1C(C(C2)O)(C)O)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 52°C to 60°C
Color White to Beige
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 2.5 g
Beilstein 06,4145
Specific Rotation Condition +8.50 (20.00°C c=6.5,toluene)
Specific Rotation + 8.5
Formula Weight 170.25
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Crystalline Mass
Chemical Name or Material (1S,2S,3R,5S)-(+)-2,3-Pinanediol
Show More Show Less

RUO – Research Use Only