missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(+)-2-Amino-1-phenyl-1,3-propanediol, 97%

Catalog No. AA4218303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4218303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4218303 Supplier Thermo Scientific Chemicals Supplier No. 04218303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Asymetric synthesis of chiral 2-oxazolines | CAS: 28143-91-1 | C9H13NO2 | 167.208 g/mol

Asymetric synthesis of chiral 2-oxazolines

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28143-91-1
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD00004503
InChI Key JUCGVCVPNPBJIG-IUCAKERBSA-N
PubChem CID 119881
IUPAC Name (1S,2S)-2-amino-1-phenylpropane-1,3-diol
SMILES C1=CC=C(C=C1)C(C(CO)N)O

Specifications

Melting Point 112°C to 115°C
Quantity 1 g
Synonym 1s,2s-+-2-amino-1-phenyl-1,3-propanediol, 1s,2s-2-amino-1-phenylpropane-1,3-diol, l-+-threo-2-amino-1-phenyl-1,3-propanediol, 1s,2s-2-amino-1-phenyl-1,3-propanediol, 1s,2s-2-amino-1-phenyl-propane-1,3-diol, 1s, 2s-+-2-amino-1-phenyl-1,3-propanediol, 1,3-propanediol, 2-amino-1-phenyl-, 1s,2s, pubchem6415, s r*,r*-2-amino-1-phenylpropane-1,3-diol, s,s-2-amino-1-phenyl-1,3-propanediol
Formula Weight 167.21
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material (1S,2S)-(+)-2-Amino-1-phenyl-1,3-propanediol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 248-867-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only