missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide, 98%

Catalog No. p-7048058
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL20170MD 250 mg
AAL2017003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL20170MD Supplier Thermo Scientific Chemicals Supplier No. L20170MD
Only null left

CAS: 132747-20-7 | C5H12Br2N2 | 259.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132747-20-7
Molecular Formula C5H12Br2N2
Molecular Weight (g/mol) 259.97
MDL Number MFCD08272741
InChI Key ISYQWKOXKGJREA-UHFFFAOYNA-N
Synonym 1s,4s-2,5-diazabicyclo 2.2.1 heptane dihydrobromide, 1s,4s-+-2,5-diazabicyclo 2.2.1 heptane dihydrobromide, 1s,4s-2,5-diazabicylo 2.2.1 heptane dihydrobromide, 2,5-diazabicyclo 2.2.1 heptane,hydrobromide 1:2, 1s,4s-2,5-diazabicyclo 2.2.1 heptane dihydrobrom, 1s,4s-2,5-diaza-bicyclo 2.2.1 heptane dihydrobromide, 1s,2s-2,5-diazabicyclo 2.2.1 heptane dihydrobromide
PubChem CID 14612497
SMILES Br.Br.C1NC2CNC1C2

Specifications

Melting Point ∽290°C (decomposition)
Quantity 250 mg
Beilstein 4258260
Sensitivity Hygroscopic
Optical Rotation +22° (c=1 in Water)
IUPAC Name 2,5-diazabicyclo[2.2.1]heptane dihydrobromide
Formula Weight 259.97
Percent Purity 98%
Chemical Name or Material (1S,2S)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only