missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(-)-1,2-Diphenylethylenediamine 98.0+%, TCI America™

Catalog No. D21751G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21751G 1 g
D21755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21751G Supplier TCI America Supplier No. D21751G

Chemical Identifiers

CAS 29841-69-8
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.296
MDL Number MFCD00082751
InChI Key PONXTPCRRASWKW-KBPBESRZSA-N
Synonym 1s,2s---1,2-diphenylethylenediamine, 1s,2s---1,2-diphenyl-1,2-ethanediamine, 1s,2s-1,2-diphenylethane-1,2-diamine, 1s,2s-1,2-diphenyl-1,2-ethanediamine, 1s,2s---1,2-diamino-1,2-diphenylethane, s,s-dpen, 1,2-ethanediamine, 1,2-diphenyl-, 1s,2s, 1s,2s---1,2-diphenyl-1,2-ethane diamine, --1,2-diphenylethylenediamine, diphenylethylenediamine
PubChem CID 6931238
IUPAC Name (1S,2S)-1,2-diphenylethane-1,2-diamine
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 1 g
Formula Weight 212.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1S,2S)-(-)-1,2-Diphenylethylenediamine