missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine, 97%

Catalog No. AAL1296803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1296803 1 g
AAL1296806 5 g
AAL1296814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1296803 Supplier Thermo Scientific Chemicals Supplier No. L1296803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 29841-69-8 | C14H16N2 | 212.296 g/mol

(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine solvation agent, For synthesis of enantiopure ethylenediamines by chirality transfer (condensation with diketones followed by reductive cleavage), Co-catalyst in the Ru catalyzed enantioselective hydrogenation of aromatic ketones, Versatile ligand for the formation of metal complexes.1 Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine solvation agent, For synthesis of enantiopure ethylenediamines by chirality transfer (condensation with diketones followed by reductive cleavage), Co-catalyst in the Ru catalyzed enantioselective hydrogenation of aromatic ketones, Versatile ligand for the formation of metal complexes.1 Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to air.

Chemical Identifiers

CAS 29841-69-8
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.296
MDL Number MFCD00082751
InChI Key PONXTPCRRASWKW-KBPBESRZSA-N
Synonym 1s,2s---1,2-diphenylethylenediamine, 1s,2s---1,2-diphenyl-1,2-ethanediamine, 1s,2s-1,2-diphenylethane-1,2-diamine, 1s,2s-1,2-diphenyl-1,2-ethanediamine, 1s,2s---1,2-diamino-1,2-diphenylethane, s,s-dpen, 1,2-ethanediamine, 1,2-diphenyl-, 1s,2s, 1s,2s---1,2-diphenyl-1,2-ethane diamine, --1,2-diphenylethylenediamine, diphenylethylenediamine
PubChem CID 6931238
IUPAC Name (1S,2S)-1,2-diphenylethane-1,2-diamine
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N

Specifications

Melting Point 80°C to 84°C
Quantity 1 g
UN Number UN3259
Beilstein 3201645
Sensitivity Air sensitive
Solubility Information Insoluble in water.
Optical Rotation −102° (c=1 in Methanol)
Formula Weight 212.3
Percent Purity 97%
Chemical Name or Material (1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine
Hazard Category H302+H312-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only