missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(-)-1,2-Cyclohexanediamine D-Tartrate 98.0+%, TCI America™

Catalog No. c14505g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C14505G 5 g
C145025G 25 g
2 options

Catalog No. C14505G

Supplier: TCI America C14505G

Only null left

Chemical Identifiers

CAS 67333-70-4
Molecular Formula C10H20N2O6
Molecular Weight (g/mol) 264.278
MDL Number MFCD00191980
InChI Key GDOTUTAQOJUZOF-SCDVTJNCSA-N
Synonym 1s,2s---1,2-diaminocyclohexane d-tartrate, 1s,2s-+-cyclohexane-1,2-diamine d-tartrate salt, 1s,2s---1,2-cyclohexanediamine d-tartrate, 1s,2s---1,2-diaminocyclohexane l-tartrate, 1s,2s-cyclohexane-1,2-diamine 2s,3s-2,3-dihydroxysuccinate, 1s,2s---cyclohexane-1,2-diamine d-tartrate, 1s,2s-1,2-diaminocyclohexane d-tartrate, 1s,2s---1,2-diaminocyclohexane-d-tartrate, 1s,2s-cyclohexane-1,2-diamine; 2s,3s-2,3-dihydroxybutanedioic acid, 1,2-cyclohexanediamine, 1s,2s-, 2s,3s-2,3-dihydroxybutanedioate 1:1
PubChem CID 16212699
IUPAC Name (1S,2S)-cyclohexane-1,2-diamine;(2S,3S)-2,3-dihydroxybutanedioic acid
SMILES C1CCC(C(C1)N)N.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Melting Point 284°C
Color White
Quantity 5 g
Formula Weight 264.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (1S,2S)-(-)-1,2-Cyclohexanediamine D-Tartrate