missing translation for 'onlineSavingsMsg'
Learn More

(1S,2R,5S)-(+)-Menthyl (R)-p-Toluenesulfinate 97.0+%, TCI America™

Supplier:  TCI America M10665G

 View more versions of this product

Catalog No. M10665G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(1S,2R,5S)-(+)-Menthyl (R)-p-Toluenesulfinate
103°C
C17H26O2S
5 g
CC(C)C1CCC(C)CC1OS(=O)C1=CC=C(C)C=C1
294.45
294.45
Crystalline Powder
91796-57-5
White
MFCD00064472,MFCD00010192
NQICGNSARVCSGJ-UHFFFAOYNA-N
5-methyl-2-(propan-2-yl)cyclohexyl 4-methylbenzene-1-sulfinate
101598140
≥97.0% (HPLC)

Chemical Identifiers

91796-57-5
294.45
NQICGNSARVCSGJ-UHFFFAOYNA-N
5-methyl-2-(propan-2-yl)cyclohexyl 4-methylbenzene-1-sulfinate
C17H26O2S
MFCD00064472,MFCD00010192
101598140
CC(C)C1CCC(C)CC1OS(=O)C1=CC=C(C)C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions