missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,2R,5S)-(+)-Menthyl (R)-p-Toluenesulfinate 97.0+%, TCI America™
Supplier: TCI America M10661G
Specifications
| (1S,2R,5S)-(+)-Menthyl (R)-p-Toluenesulfinate | |
| 103°C | |
| C17H26O2S | |
| 1 g | |
| CC(C)C1CCC(C)CC1OS(=O)C1=CC=C(C)C=C1 | |
| 294.45 | |
| 294.45 | |
| Crystalline Powder |
| 91796-57-5 | |
| White | |
| MFCD00064472,MFCD00010192 | |
| NQICGNSARVCSGJ-UHFFFAOYNA-N | |
| 5-methyl-2-(propan-2-yl)cyclohexyl 4-methylbenzene-1-sulfinate | |
| 101598140 | |
| ≥97.0% (HPLC) |
Chemical Identifiers
| 91796-57-5 | |
| 294.45 | |
| NQICGNSARVCSGJ-UHFFFAOYNA-N | |
| 5-methyl-2-(propan-2-yl)cyclohexyl 4-methylbenzene-1-sulfinate |
| C17H26O2S | |
| MFCD00064472,MFCD00010192 | |
| 101598140 | |
| CC(C)C1CCC(C)CC1OS(=O)C1=CC=C(C)C=C1 |
Safety and Handling
TSCA : No
Recommended Storage : Freezer