missing translation for 'onlineSavingsMsg'
Learn More

(1S,2R)-(+)-2-Amino-1,2-diphenylethanol 99.0+%, TCI America™

Catalog No. p-7117320
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12311G 1 g
A12315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12311G Supplier TCI America Supplier No. A12311G
Only null left

Chemical Identifiers

CAS 23364-44-5
Molecular Formula C14H15NO
Molecular Weight (g/mol) 213.28
MDL Number MFCD00074959
InChI Key GEJJWYZZKKKSEV-KGLIPLIRSA-N
Synonym 1s,2r-2-amino-1,2-diphenylethanol, 1s,2r-+-2-amino-1,2-diphenylethanol, 1s,2r-2-amino-1,2-diphenyl-ethanol, 1s,2r-2-amino-1,2-diphenylethan-1-ol, pubchem5988, gejjwyzzkkksev-kgliplirsa, 1s,2r-2-amino-1,2-di phenyl ethanol, 1r,2s-1-amino-1,2-diphenyl-2-ethanol;, 1s,2r-+-erythro-2-amino-1,2-diphenylethanol, racemic-1s,2r-2-amino-1,2-diphenylethan-1-ol
PubChem CID 719822
IUPAC Name (1S,2R)-2-amino-1,2-diphenylethanol
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)N

Specifications

Melting Point 144°C
Color White
Quantity 1 g
Formula Weight 213.28
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1S,2R)-(+)-2-Amino-1,2-diphenylethanol