missing translation for 'onlineSavingsMsg'
Learn More

(1R)-endo-(+)-Fenchyl alcohol, 96%

Catalog No. AAL0321136
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0321136 500 g
AAL0321122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0321136 Supplier Thermo Scientific Chemicals Supplier No. L0321136

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2217-02-9 | C4H4N2O5 | 160.085 g/mol

(1R)-endo-(+)-Fenchyl alcohol can be extensively used in daily chemicals such as fair antiperspirant, textile softener and liquid detergent. It can also be esterified by different kinds of organic acids to expand its usage in flavors and fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R)-endo-(+)-Fenchyl alcohol can be extensively used in daily chemicals such as fair antiperspirant, textile softener and liquid detergent. It can also be esterified by different kinds of organic acids to expand its usage in flavors and fragrances.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Incompatible with oxidizing agents. Ensure good ventilation.

Chemical Identifiers

CAS 2217-02-9
MDL Number MFCD00003760

Specifications

Melting Point 37°C to 44°C
Boiling Point 201°C to 202°C
Quantity 500 g
Flash Point 73°C (163°F)
Odor Camphor-like
Optical Rotation +10.5° (c = 10 in Ethanol)
Molecular Formula C4H4N2O5
Beilstein 1850917
Synonym (+)-Fenchol; 1,3,3-Trimethyl-2-norbornanol
Solubility Information Insoluble in water.
InChI Key IAIHUHQCLTYTSF-MRTMQBJTSA-N
SMILES CC1(C2CCC(C2)(C1O)C)C
IUPAC Name (1S,3R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-ol
Molecular Weight (g/mol) 154.25
PubChem CID 6997371
Formula Weight 154.25
Percent Purity 96%
Chemical Name or Material (1R)-endo-(+)-Fenchyl alcohol
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 216-639-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only