missing translation for 'onlineSavingsMsg'
Learn More

(1R,E)-(+)-Camphorquinone 3-oxime, 99%, Thermo Scientific Chemicals

Catalog No. AAH5103406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5103406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5103406 Supplier Thermo Scientific Chemicals Supplier No. H5103406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 31571-14-9 | Molecular Formula: C10H15NO2 | Molecular Weight: 181.235 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31571-14-9
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.235
MDL Number MFCD00074848
InChI Key YRNPDSREMSMKIY-WKEGUHRASA-N
Synonym anti-1r-+-camphorquinone3-oxime, 1s,4r-2-hydroxyimino-4,7,7-trimethylbicyclo 2.2.1 heptan-3-one
PubChem CID 23308125
IUPAC Name (1R,4R)-2-hydroxyimino-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C2CCC1(C(=O)C2=NO)C)C

Specifications

Quantity 5 g
Beilstein 3200377
Formula Weight 181.23
Percent Purity 99%
Chemical Name or Material (1R,E)-(+)-Camphorquinone 3-oxime
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only