missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R)-Camphor Oxime 98.0+%, TCI America™
$52.19 - $464.14
Chemical Identifiers
| CAS | 2792-42-9 |
|---|---|
| Molecular Formula | C10H17NO |
| Molecular Weight (g/mol) | 167.25 |
| MDL Number | MFCD00046298,MFCD00210161 |
| InChI Key | OVFDEGGJFJECAT-DHZHZOJONA-N |
| PubChem CID | 124925274 |
| IUPAC Name | N-[(2E)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ylidene]hydroxylamine |
| SMILES | CC1(C)C2CCC1(C)\C(C2)=N\O |
Chemical Identifiers
| 2792-42-9 | |
| 167.25 | |
| OVFDEGGJFJECAT-DHZHZOJONA-N | |
| N-[(2E)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ylidene]hydroxylamine |
| C10H17NO | |
| MFCD00046298,MFCD00210161 | |
| 124925274 | |
| CC1(C)C2CCC1(C)\C(C2)=N\O |
Specifications
| 2792-42-9 | |
| White | |
| MFCD00046298,MFCD00210161 | |
| OVFDEGGJFJECAT-DHZHZOJONA-N | |
| N-[(2E)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ylidene]hydroxylamine | |
| 124925274 | |
| ≥98.0% (GC) | |
| (1R)-Camphor Oxime |
| 119°C | |
| C10H17NO | |
| 1 g | |
| CC1(C)C2CCC1(C)\C(C2)=N\O | |
| 167.25 | |
| 167.25 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes