missing translation for 'onlineSavingsMsg'
Learn More

(1R)-(-)-Camphor-10-sulfonic acid, 98%

Catalog No. AAB2155336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2155336 500 g
AAB2155314 25 g
AAB2155322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2155336 Supplier Thermo Scientific Chemicals Supplier No. B2155336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 35963-20-3 | C10H16O4S | 232.29 g/mol

(1R)-(-)-Camphor-10-sulfonic acid, is used as a pharamaceutical intermediate and also used as a chiral derivative of Camp. It can also be used as resolving agents for chiral amines and other cations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R)-(-)-Camphor-10-sulfonic acid, is used as a pharamaceutical intermediate and also used as a chiral derivative of Camp. It can also be used as resolving agents for chiral amines and other cations.

Solubility
Soluble in water, and a wide variety of organic substances.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 35963-20-3
Molecular Formula C10H16O4S
Molecular Weight (g/mol) 232.29
MDL Number MFCD00150753,MFCD00064158
InChI Key MIOPJNTWMNEORI-MHPPCMCBSA-N
Synonym l--camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, 1r---camphor-10-sulfonic acid, unii-y6075i4fxe, camphorsulfonic acid,-, l---camphor-10-sulfonic acid, s-camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 hept-1-yl methanesulfonic acid, --camphor-10-sulfonic acid
PubChem CID 5771688
ChEBI CHEBI:55401
SMILES CC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2

Specifications

Melting Point ∼200°C
Quantity 500 g
UN Number UN3261
Beilstein 2809676
Sensitivity Hygroscopic
Solubility Information Soluble in water,and a wide variety of organic substances.
Optical Rotation −21° (c=10 in Water)
IUPAC Name [(1S,4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
Formula Weight 232.3
Percent Purity ≥98%
Chemical Name or Material (1R)-(-)-Camphor-10-sulfonic acid
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 252-817-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only