missing translation for 'onlineSavingsMsg'
Learn More

(1R,3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee

Catalog No. p-70147
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC361832500 250 mg Glass Bottle
AC361830010 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC361832500 Supplier Thermo Scientific Chemicals Supplier No. 361832500
Only null left

CAS: 261165-05-3 | C11H19NO4 | 229.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 261165-05-3
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.27
MDL Number MFCD01320857
InChI Key RNJQBGXOSAQQDG-JGVFFNPUSA-N
Synonym 1s,3r-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, +-1s,3r-n-boc-3-aminocyclopentanecarboxylic acid, 1r,3s-n-boc-1-aminocyclopentane-3-carboxylic acid, 1s,3r-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, cis-3-boc-amino cyclopentane-1-carboxylic acid, 1s,3r-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1s,3r-3-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid
PubChem CID 1512529
IUPAC Name (1S,3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCC(C1)C(=O)O

Specifications

Melting Point 111.8°C
CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min. (HPLC)
Assay Percent Range 94% min. (ELSD) (HPLC)
Packaging Glass Bottle
Quantity 250 mg
Solubility Information Solubility in water: soluble
Formula Weight 229.27
Percent Purity 95%,98% ee
Physical Form Powder and/or Chunks
Chemical Name or Material (1R, 3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only