missing translation for 'onlineSavingsMsg'
Learn More

(1R,3S)-(-)-3-(Boc-amino)cyclopentanecarboxylic acid, 95%, Thermo Scientific Chemicals

Catalog No. AAH5202203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5202203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5202203 Supplier Thermo Scientific Chemicals Supplier No. H5202203

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 161660-94-2
Molecular Formula C11H18NO4
Molecular Weight (g/mol) 228.27
MDL Number MFCD01320858,MFCD01320857
InChI Key RNJQBGXOSAQQDG-SFYZADRCSA-M
Synonym 1r,3s-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, --1r,3s-n-boc-3-aminocyclopentanecarboxylic acid, cis-3-boc-amino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3s-boc-3-aminocyclopentane-1-carboxylic acid, cis-3-boc-amino cyclopentane-1-carboxylic acid, --1r,3s-n-boc-3-aminocyclopentane carboxylic acid
PubChem CID 1512526
SMILES CC(C)(C)OC(=O)N[C@H]1CC[C@H](C1)C([O-])=O

Specifications

IUPAC Name (1R,3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
Quantity 1 g
Formula Weight 229.28
Percent Purity 95%
Chemical Name or Material (1R,3S)-(-)-3-(Boc-amino)cyclopentanecarboxylic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only