missing translation for 'onlineSavingsMsg'
Learn More

(1R,3R)-5-(Bromomethylene)-1,3-bis(tert-butyldimethylsilyloxy)cyclohexane 95.0+%, TCI America™

Catalog No. B2779100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2779100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B2779100MG Supplier TCI America Supplier No. B2779100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 387834-41-5
Molecular Formula C19H39BrO2Si2
Molecular Weight (g/mol) 435.593
InChI Key GCYQQWTZQGQJBW-IAGOWNOFSA-N
PubChem CID 10137871
IUPAC Name [(1R,5R)-3-(bromomethylidene)-5-[tert-butyl(dimethyl)silyl]oxycyclohexyl]oxy-tert-butyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OC1CC(CC(=CBr)C1)O[Si](C)(C)C(C)(C)C

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 435.59
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (1R,3R)-5-(Bromomethylene)-1,3-bis(tert-butyldimethylsilyloxy)cyclohexane