missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine 98.0+%, TCI America™

$426.87 - $1461.41

Chemical Identifiers

CAS 364385-54-6
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD09952106
InChI Key AKVIZYGPJIWKOS-DTWKUNHWSA-N
Synonym (1R,2S)-N1-Boc-1,2-cyclohexanediamine, (1R,2S)-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane, (1R,2S)-2-Amino-1-(Boc-amino)cyclohexane
PubChem CID 1514392
IUPAC Name tert-butyl N-[(1R,2S)-2-aminocyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCCC1N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B34781G
View Documents
TCI America
B34781G
1 g
Each for $426.87
Only null left
 
B34785G
View Documents
TCI America
B34785G
5 g
Each for $1,461.41
Only null left
 
Description

Description

Specifications

Chemical Identifiers

364385-54-6
214.309
AKVIZYGPJIWKOS-DTWKUNHWSA-N
1514392
CC(C)(C)OC(=O)NC1CCCCC1N
C11H22N2O2
MFCD09952106
(1R,2S)-N1-Boc-1,2-cyclohexanediamine, (1R,2S)-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane, (1R,2S)-2-Amino-1-(Boc-amino)cyclohexane
tert-butyl N-[(1R,2S)-2-aminocyclohexyl]carbamate

Specifications

364385-54-6
C11H22N2O2
1 g
AKVIZYGPJIWKOS-DTWKUNHWSA-N
tert-butyl N-[(1R,2S)-2-aminocyclohexyl]carbamate
1514392
≥98.0% (GC,T)
(1R,2S)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine
White
MFCD09952106
(1R,2S)-N1-Boc-1,2-cyclohexanediamine, (1R,2S)-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane, (1R,2S)-2-Amino-1-(Boc-amino)cyclohexane
CC(C)(C)OC(=O)NC1CCCCC1N
214.309
214.31
Crystalline Lumps
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'