missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S,5R)-(-)-Menthyl (S)-p-toluenesulfinate, 98%

Catalog No. AC428110100
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428110100 10 g Glass bottle
AC428110010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428110100 Supplier Thermo Scientific Chemicals Supplier No. 428110100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1517-82-4 | C17H26O2S | 294.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1517-82-4
Molecular Formula C17H26O2S
Molecular Weight (g/mol) 294.46
InChI Key NQICGNSARVCSGJ-DSRNLFJRSA-N
Synonym 1r,2s,5r---menthyl s-p-toluenesulfinate, 4-methylbenzenesulfinic acid 1r,5r-2-isopropyl-5-methylcyclohexyl ester
PubChem CID 101348873
IUPAC Name [(1R,5R)-5-methyl-2-propan-2-ylcyclohexyl] 4-methylbenzenesulfinate
SMILES CC1CCC(C(C1)OS(=O)C2=CC=C(C=C2)C)C(C)C

Specifications

Melting Point 102.0°C to 104.0°C
Color Brown to Yellow or Gray
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Specific Rotation −190° to −204° (20°C, 589nm) (c=2, acetone)
Formula Weight 294.46
Percent Purity 98%
Physical Form Solution
Chemical Name or Material (1R, 2S, 5R)-(-)-Menthyl (S)-p-toluenesulfinate
Show More Show Less

RUO – Research Use Only