missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S)-(-)-2-Amino-1,2-diphenylethanol 99.0+%, TCI America™

Catalog No. A12305G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12305G 5 g
A12301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12305G Supplier TCI America Supplier No. A12305G

Chemical Identifiers

CAS 23190-16-1
Molecular Formula C14H15NO
Molecular Weight (g/mol) 213.28
MDL Number MFCD00074960
InChI Key GEJJWYZZKKKSEV-UONOGXRCSA-N
Synonym 1r,2s-2-amino-1,2-diphenylethanol, 1r,2s---2-amino-1,2-diphenylethanol, 1r,2s-2-amino-1,2-diphenyl-ethanol, 1r,2s-2-amino-1,2-diphenylethan-1-ol, 1s,2r-1,2-diphenyl-2-hydroxyethylamine, pubchem5986, ksc201s9n, gejjwyzzkkksev-uonogxrcsa, erythro-2-amino-1,2-diphenylethanol, 1r,2s---diphenyl-2-aminoethanol
PubChem CID 719819
IUPAC Name (1R,2S)-2-amino-1,2-diphenylethanol
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)N

Specifications

Melting Point 144°C
Quantity 5 g
Formula Weight 213.28
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2S)-(-)-2-Amino-1,2-diphenylethanol